AC5410538032 Accela ChemBio Inc.

3-Acetamidobenzylamine

  • MDL number MFCD05669728
  • Cas Number : 96783-68-5
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025408&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025408.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart