AC5410538035 Accela ChemBio Inc.

5-Fluoro-2-methyl-4-nitropyridine 1-Oxide

  • MDL number MFCD12828024
  • Cas Number : 113209-88-4
  • Molecular Weight : 172.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025429&attname=GoodsCode

Safety Information

signalword Warning
MSDS http://pdf.accelachem.com/cn/msds/SY025429.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart