AC5410538036 Accela ChemBio Inc.

1-Bromo-2-fluoro-5-methoxy-4-nitrobenzene

  • MDL number MFCD16658620
  • Cas Number : 330794-02-0
  • Molecular Weight : 250.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025432&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025432.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart