AC5410538038 Accela ChemBio Inc.

5-Amino-2-(hydroxymethyl)pyridine

  • MDL number MFCD07368188
  • Cas Number : 873651-92-4
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025459&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025459.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart