AC5410538039 Accela ChemBio Inc.

2-Bromo-5-fluorobenzylamine

  • MDL number MFCD06212850
  • Cas Number : 747392-34-3
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025505&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY025505.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart