AC5410538042 Accela ChemBio Inc.

Ethyl 2-Methyl-3-furoate

  • MDL number MFCD00134327
  • Cas Number : 28921-35-9
  • Molecular Weight : 154.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025556&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P280-P370+P378-P403+P235-P501
hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY025556.pdf
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart