AC5410538043 Accela ChemBio Inc.

4-Bromo-2-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD23705605
  • Cas Number : 63272-66-2
  • Molecular Weight : 231.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025557&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025557.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart