AC5410538050 Accela ChemBio Inc.

2-Amino-6-ethylbenzoic Acid

  • MDL number MFCD11616258
  • Cas Number : 66232-56-2
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025652&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025652.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart