AC5410538059 Accela ChemBio Inc.

Methyl 3-Formyl-5-methoxybenzoate

  • MDL number MFCD11045770
  • Cas Number : 69026-10-4
  • Molecular Weight : 194.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025806&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025806.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart