AC5410538065 Accela ChemBio Inc.

N-Acetyl-D-allo-isoleucine

  • MDL number MFCD00136538
  • Cas Number : 54831-20-8
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025916&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025916.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart