AC5410538073 Accela ChemBio Inc.

Dimethyl 4-Nitroisophthalate

  • MDL number MFCD00177948
  • Cas Number : 69048-70-0
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025954&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025954.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart