AC5410538077 Accela ChemBio Inc.

5-Amino-2-(Boc-aminomethyl)pyridine

  • MDL number MFCD09839773
  • Cas Number : 1780934-03-3
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025993&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025993.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart