AC5410538095 Accela ChemBio Inc.

5-(Aminomethyl)-2-fluoroanisole

  • MDL number MFCD04116362
  • Cas Number : 508177-67-1
  • Molecular Weight : 155.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026389&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY026389.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart