AC5410538100 Accela ChemBio Inc.

3-(Hydroxymethyl)-2-methylfuran

  • MDL number MFCD00052572
  • Cas Number : 5554-99-4
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026412&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302+P352-P261-P301+P312-P304+P340-P280-P305+P351+P338
MSDS http://pdf.accelachem.com/cn/msds/SY026412.pdf
pictograms GHS07
hstatements H332-H315-H335-H302-H319-H312
Availability details for
Quantity
  • Availability
  • Add to Cart