AC5410538101 Accela ChemBio Inc.

1-Benzhydryl-3-chloroazetidine

  • MDL number MFCD11519007
  • Cas Number : 959256-87-2
  • Molecular Weight : 257.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026422&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026422.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart