AC5414275451 Accela ChemBio Inc.

Diethyl 5-Bromopyrimidine-4,6-dicarboxylate

  • MDL number MFCD26937564
  • Cas Number : 1820687-49-7
  • Molecular Weight : 303.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026892&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026892.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart