AC5414275452 Accela ChemBio Inc.

2-(Allyloxy)-4,6-dimethoxybenzyl Alcohol

  • MDL number MFCD27500644
  • Cas Number : 1820712-30-8
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027370&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027370.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart