AC5414275453 Accela ChemBio Inc.

2-(Allyloxy)-6-bromo-4-methoxybenzyl Alcohol

  • MDL number MFCD27500645
  • Cas Number : 1820705-12-1
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027371&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027371.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart