AC5414275455 Accela ChemBio Inc.

4,4'-[1,4-Phenylenebis(ethyne-2,1-diyl)]dibenzoic Acid

  • MDL number MFCD27500652
  • Cas Number : 217077-89-9
  • Molecular Weight : 366.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027717&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027717.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart