AC5414275457 Accela ChemBio Inc.

Ethyl 5-[4-(Ethoxycarbonyl)phenyl]isoxazole-3-carboxylate

  • MDL number MFCD27935387
  • Cas Number : 1820686-51-8
  • Molecular Weight : 289.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028061&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028061.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart