AC5414275458 Accela ChemBio Inc.

Methyl 3-(Boc-amino)-3-(3-hydroxyphenyl)propanoate

  • MDL number MFCD27935388
  • Cas Number : 1423134-61-5
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028065&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028065.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart