AC5414275459 Accela ChemBio Inc.

3-Amino-4,5,6,7-tetrahydro-1H-indazole Hydrochloride

  • MDL number MFCD28053752
  • Cas Number : 1376043-30-9
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028241&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY028241.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart