AC5414275493 Accela ChemBio Inc.

Benzyl N-Boc-4-isopropyl-4-piperidinecarboxylate

  • MDL number MFCD16495869
  • Cas Number : 1226776-80-2
  • Molecular Weight : 361.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007986&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY007986.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart