AC5414275501 Accela ChemBio Inc.

4,5,6,7-Tetrahydro-1,2,3-triazolo[1,5-a]pyrazine

  • MDL number MFCD09802206
  • Cas Number : 123291-54-3
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008689&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY008689.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart