AC5414275508 Accela ChemBio Inc.

3-Amino-4,5,6,7-tetrahydro-1H-indazole

  • MDL number MFCD12827628
  • Cas Number : 55440-17-0
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011633&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011633.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart