AC5414275512 Accela ChemBio Inc.

3-(Boc-amino)-6-fluoro-1H-indazole

  • MDL number MFCD18651681
  • Cas Number : 1176089-41-0
  • Molecular Weight : 251.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012349&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012349.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart