AC5414275513 Accela ChemBio Inc.

1-Benzoyl-6-bromo-7-azaindole

  • MDL number MFCD09880008
  • Cas Number : 143468-12-6
  • Molecular Weight : 301.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012372&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012372.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart