AC5414275514 Accela ChemBio Inc.

1-Methyl-2-oxopyrrolidine-4-carboxamidine Hydrochloride

  • MDL number MFCD18651696
  • Cas Number : 1272756-18-9
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012399&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012399.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart