AC5414275518 Accela ChemBio Inc.

Ethyl 2-(5-Oxazolyl)benzoate

  • MDL number MFCD18375300
  • Cas Number : 1186127-15-0
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012500&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012500.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart