AC5414275520 Accela ChemBio Inc.

2-[3-(2-Amino-4-thiazolyl)phenoxy]acetic Acid

  • MDL number MFCD11564607
  • Cas Number : 1082128-37-7
  • Molecular Weight : 250.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012615&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012615.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart