AC5414275527 Accela ChemBio Inc.

Ethyl 4-Benzyl-2-morpholinecarboxylate Hydrochloride

  • MDL number MFCD18911733
  • Cas Number : 135782-26-2
  • Molecular Weight : 285.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013594&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013594.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart