AC5414275534 Accela ChemBio Inc.

4-Amino-1-(methoxymethyl)pyrazole

  • MDL number MFCD04969823
  • Cas Number : 1001500-80-6
  • Molecular Weight : 127.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014285&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014285.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart