AC5414275535 Accela ChemBio Inc.

(3-Bromophenyl)(2-thienyl)methanone

  • MDL number MFCD07658070
  • Cas Number : 31161-47-4
  • Molecular Weight : 267.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014491&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014491.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart