AC5414275540 Accela ChemBio Inc.

5-(4-Carboxyphenyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD22192330
  • Cas Number : 1375064-71-3
  • Molecular Weight : 233.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015169&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015169.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart