AC5414275541 Accela ChemBio Inc.

4-[3-(Methoxycarbonyl)-5-isoxazolyl]benzoic Acid

  • MDL number MFCD22192331
  • Cas Number : 1375064-52-0
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015170&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015170.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart