AC5414275545 Accela ChemBio Inc.

5-(2-Pyrazinyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD22192339
  • Cas Number : 1375064-55-3
  • Molecular Weight : 191.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015185&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015185.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart