AC5414275552 Accela ChemBio Inc.

2-(Boc-amino)-2-(3,4-dichlorophenyl)acetic Acid

  • MDL number MFCD03425232
  • Cas Number : 142121-95-7
  • Molecular Weight : 320.1685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015955&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015955.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart