AC5414275555 Accela ChemBio Inc.

Sodium 2-Methoxy-2-oxoethanesulfonate

  • MDL number MFCD11044485
  • Cas Number : 29508-16-5
  • Molecular Weight : 176.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016112&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016112.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart