AC5414275557 Accela ChemBio Inc.

6-(2-Chlorophenyl)-3-pyridinecarbaldehyde

  • MDL number MFCD04115437
  • Cas Number : 898404-60-9
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016238&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016238.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart