AC5414275576 Accela ChemBio Inc.

Methyl 5-Cyclopropylisoxazole-3-carboxylate

  • MDL number MFCD07390343
  • Cas Number : 887360-91-0
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018272&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018272.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart