AC5414275584 Accela ChemBio Inc.

1-Cbz-3-ethylpiperidine-3-carboxylic Acid

  • MDL number MFCD17976788
  • Cas Number : 1245808-57-4
  • Molecular Weight : 291.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018688&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018688.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart