AC5414275585 Accela ChemBio Inc.

1-Boc-3-isopropylpiperidine-3-carboxylic Acid

  • MDL number MFCD21641317
  • Cas Number : 1363165-91-6
  • Molecular Weight : 271.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018691&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018691.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart