AC5414275586 Accela ChemBio Inc.

Ethyl 1-Cbz-3-isopropylpiperidine-3-carboxylate

  • MDL number MFCD21641318
  • Cas Number : 1363166-26-0
  • Molecular Weight : 333.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018692&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018692.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart