AC5414275588 Accela ChemBio Inc.

2,4-Dimethylbenzoxazole

  • MDL number MFCD21641329
  • Cas Number : 72692-90-1
  • Molecular Weight : 147.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018726&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018726.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart