AC5414275602 Accela ChemBio Inc.

alpha-Acetoxyphenylacetonitrile

  • MDL number MFCD22205885
  • Cas Number : 5762-35-6
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019891&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019891.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart