AC5414275606 Accela ChemBio Inc.

4-Nitro-1,2,3-triazole

  • MDL number MFCD00730971
  • Cas Number : 84406-63-3
  • Molecular Weight : 114.06288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020363&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY020363.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart