AC5414275613 Accela ChemBio Inc.

8-Methyl-2(1H)-quinolinone

  • MDL number MFCD13195790
  • Cas Number : 4053-36-5
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-methyl-2(1H)-quinolinone
Inchi Code 1S/C10H9NO/c1-7-3-2-4-8-5-6-9(12)11-10(7)8/h2-6H,1H3,(H,11,12)
Inchi Key XXPVCQMOIBCSDT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to orange solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021046&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021046.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart