AC5414275614 Accela ChemBio Inc.

2-Amino-4-hydroxypyridine Hydrochloride

  • MDL number MFCD09701349
  • Cas Number : 1187932-09-7
  • Molecular Weight : 146.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-pyridinol hydrochloride
Inchi Code 1S/C5H6N2O.ClH/c6-5-3-4(8)1-2-7-5;/h1-3H,(H3,6,7,8);1H
Inchi Key DEVWTUWLHTYPTL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021111&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021111.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart