AC5414275624 Accela ChemBio Inc.

(2,2-Dibromovinyl)benzene

  • MDL number MFCD01024288
  • Cas Number : 7436-90-0
  • Molecular Weight : 261.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022051&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022051.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart