AC5414275631 Accela ChemBio Inc.

Ethyl 3-(4-Pyridyl)propiolate

  • MDL number MFCD23381103
  • Cas Number : 66869-71-4
  • Molecular Weight : 175.18396

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022415&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY022415.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart